3-(2-Thienyl)-D-alanine

Supplier: BACHEM AMERICAS INC

Synonyms: H-β-(2-Thienyl)-D-Ala-OH, (R)-2-Amino-3-(thiophen-2-yl)propanoic acid, H-D-3-(2-Thienyl)-Ala-OH

4008202.0001 4008202.0005
F-2115.0001BAEA 748.67 CAD
F-2115.0001BA F-2115.0005BA
3-(2-Thienyl)-D-alanine
3-(2-Thienyl)-D-alanine

Sequence: H-β-(2-Thienyl)-D-Ala-OH


2-Thi derivatives.


H-D-Thi-OH.

Formula: C₇H₉NO₂S
Storage Temperature: Ambient
MDL Number: MFCD00077166
CAS Number: 62561-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR