3-(2-Thienyl)-L-alanine

Supplier: BACHEM AMERICAS INC

Synonyms: H-β-(2-Thienyl)-Ala-OH, 3-(2-Thienyl)-L-alanine, (S)-2-Amino-3-(thiophen-2-yl)propanoic acid

4006811.0001 4006811.0005
F-2110.0001BAEA 215.16 CAD
F-2110.0001BA F-2110.0005BA
3-(2-Thienyl)-L-alanine
3-(2-Thienyl)-L-alanine

Sequence: H-β-(2-Thienyl)-Ala-OH


2-Thi derivatives.


L-β-(2-Thienyl)-alanine (H-Thi-OH) is structurally similar to L-phenylalanine. H-Thi-OH inhibits the growth of Escherichia coli, presumably by competitive inhibition of enzymes for which Phe is the substrate.

Formula: C₇H₉NO₂S
MW: 171.214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064335
CAS Number: 22951-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR