Ethyl-2-aminopropanoate

Supplier: BACHEM AMERICAS INC

Synonyms: H-DL-Ala-OEt·HCl

4011110.0025 4011110.0250
F-2095.0025BAEA 211.77 CAD
F-2095.0025BA F-2095.0250BA
Ethyl-2-aminopropanoate
Ethyl-2-aminopropanoate

Sequence: H-DL-Ala-OEt · HCl


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).


[617-27-6] (hydrochloride)

Formula: C₅H₁₁NO₂
MW: 117.148 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00013018
CAS Number: 17344-99-9
EINECS: 210-507-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR