(S)-Methyl-2-aminopent-4-ynoate hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-Methyl 2-aminopent-4-ynoate hydrochloride, H-Pra-OMe·HCl

4010420.0250 4010420.1000
F-2075.0250BAEA 335.58 CAD
F-2075.0250BA F-2075.1000BA
(S)-Methyl-2-aminopent-4-ynoate hydrochloride
(S)-Methyl-2-aminopent-4-ynoate hydrochloride

Sequence: H-Pra-OMe · HCl
Synonym(s): H-Propargyl-Gly-OMe · HCl#(S)-2-Amino-4-pentynoic acid-methyl ester · HCl


Bachem also offers resin-bound propargylglycine, Fmoc-Pra-Wang resin (D-2820).


Nα-protected H-Pra-OMe has been treated with aryl iodides/CuI/Pd(PPh₃)₄ (Sonogashira reaction) to obtain aryl-substituted propargylglycine derivatives.

Formula: C₆H₁₀ClNO₂
MW: 163.601 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00238137
CAS Number: 166271-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR