D-(+)-Proline

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Pro-OH, (R)-(+)-Proline, D-Proline

4031089.0005 4031089.0100
F-2045.0005BAEA 227.28 CAD
F-2045.0005BA F-2045.0100BA
D-(+)-Proline
D(+)-Proline

Sequence: H-D-Pro-OH

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064317
CAS Number: 344-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR