L-O-Phosphoserine

Supplier: BACHEM AMERICAS INC

4002875.0005 4002875.0025 4002875.0100
F-2030.0005BAEA 232.35 CAD
F-2030.0005BA F-2030.0025BA F-2030.0100BA
L-O-Phosphoserine

Sequence: H-Ser(PO₃H₂)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.
N-Terminal phosphoserine residues react with thioesters yielding highly reactive acylphosphates which spontaneously undergo an intramolecular O-N-shift allowing chemical ligation at such positions.


Synonym(s): Dexfosfoserine

Formula: C₃H₈NO₆P
Storage Temperature: Freezer
MDL Number: MFCD00065935
CAS Number: 407-41-0
EINECS: 206-986-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR