H-Phg-OtBu·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-Phg-OtBu.HCl, H-Phg-OtBu·HCl

4001013.0001 4001013.0005 4001013.0025
F-2020.0001BAEA 180.83 CAD
F-2020.0001BA F-2020.0005BA F-2020.0025BA
H-Phg-OtBu·HCl
H-Phg-OtBu·HCl

Sequence: H-Phg-OtBu · HCl

Formula: C₁₂H₁₈ClNO₂
Storage Temperature: Freezer
MDL Number: MFCD00153456
CAS Number: 161879-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR