D-(-)-Ornithine hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Orn-OH·HCl, H-D-Orn-OH.HCl

4031134.0005 4031134.0025 4031134.0100
F-1970.0005BAEA 180.83 CAD
F-1970.0005BA F-1970.0025BA F-1970.0100BA
D-(-)-Ornithine hydrochloride
D(-)-Ornithine hydrochloride

Sequence: H-D-Orn-OH · HCl

Formula: C₅H₁₂N₂O₂·HCl
MW: 168.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012917
CAS Number: 16682-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR