L-Norvaline

Supplier: BACHEM AMERICAS INC

4031120.0005 4031120.0025
F-1940.0005BAEA 180.91 CAD
F-1940.0005BA F-1940.0025BA
L-Norvaline
L-Norvaline

Sequence: H-Nva-OH
Synonym(s): L-2-Aminovaleric acid


Inhibitor of arginase.

Formula: C₅H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00064421
CAS Number: 6600-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR