D-(-)-Norleucine

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Nle-OH, (R)-(-)-Norleucine

4031116.0005 4031116.0025
F-1920.0005BAEA 283.94 CAD
F-1920.0005BA F-1920.0025BA
D-(-)-Norleucine
D(-)-Norleucine

Sequence: H-D-Nle-OH
Synonym(s): D-2-Aminohexanoic acid

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 301
Storage Temperature: Ambient
MDL Number: MFCD00008099
CAS Number: 327-56-0
Merck Index: 12,06800

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR