L(+)-Norleucine

Supplier: BACHEM AMERICAS INC

Synonyms: H-Nle-OH, H-NorLeu-OH, (S)-2-Aminohexanoic acid, (S)-(+)-Norleucine

4007429.0005 4007429.0025
F-1915.0005BAEA 183.09 CAD
F-1915.0005BA F-1915.0025BA
L(+)-Norleucine
L(+)-Norleucine

Sequence: H-Nle-OH
Synonym(s): L-2-Aminohexanoic acid

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064423
CAS Number: 327-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR