4-Nitro-D-phenylalanine hydrate
Supplier: BACHEM AMERICAS INC
Synonyms:
H-D-Phe(4-NO2)-OH.H2O, H-D-Phe(4-NO2)-OH·H2O, (2R)-2-Amino-3-(4-nitrophenyl)propanoic acid hydrate
Sequence: H-p-Nitro-D-Phe-OH
Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
p-Nitrophenylalanine (4-Nph, Phe(4-NO₂)) is incorporated into peptides as a chromophore. In FRET substrates, it acts as quencher. Bachem also offers resin-bound Fmoc-Nph (Fmoc-p-nitro-Phe-Wang resin, D-1135).
Formula:
C₉H₁₀N₂O₄·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150962 CAS Number: 444777-67-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...