Nω-Nitro-L-arginine methyl ester hydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride, L-NAME Hydrochloride, L-NAME HCl, H-Arg(NO2)-OMe.HCl, (S)-Methyl 2-amino-5-(3-nitroguanidino)pentanoate hydrochloride
4001301.0025
4001301.0100
F-1885.0025BAEA
309.66
CAD
F-1885.0025BA
F-1885.0100BA
Nω-Nitro-L-arginine methyl ester hydrochloride
Nomega-Nitro-L-arginine methyl ester hydrochloride
Sequence: H-Arg(NO₂)-OMe · HCl
Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).
Synonym(s): L-NAME · HCl
Formula:
C₇H₁₅N₅O₄ MW: 269.69 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00039052 CAS Number: 51298-62-5 EINECS: 257-116-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...