H-Arg(NO₂)-OBzl p-tosylate salt
Supplier: BACHEM AMERICAS INC
4007045.0005
4007045.0025
F-1880.0005BAEA
206.44
CAD
F-1880.0005BA
F-1880.0025BA
H-Arg(NO₂)-OBzl p-tosylate salt
H-Arg(NO₂)-OBzl p-tosylate salt
Sequence: H-Arg(NO₂)-OBzl p-tosylate salt
Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also F-1870, H-Arg(NO₂)-OH, in the subfamily 'Arginine Substrates and Inhibitors'.
Formula:
C₁₃H₁₉N₅O₄ MW: 309.33 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00083444 CAS Number: 7672-27-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...