α-Methyl-DL-tryptophan
Supplier: BACHEM AMERICAS INC
Synonyms:
H-α-Me-DL-Trp-OH, 2-Amino-3-(1H-indol-3-yl)-2-methylpropanoic acid
4008387.0001
4008387.0005
F-1810.0001BAEA
258.14
CAD
F-1810.0001BA
F-1810.0005BA
α-Methyl-DL-tryptophan
alpha-Methyl-DL-tryptophan
Sequence: H-α-Me-DL-Trp-OH
MeTrp, αMeTrp.
Formula:
C₁₂H₁₄N₂O₂ MW: 218.256 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038416 CAS Number: 153-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...