DL-α-Methylleucine

Supplier: BACHEM AMERICAS INC

Synonyms: H-α-Me-DL-Leu-OH

4004270.0005 4004270.0025
F-1800.0005BAEA 309.81 CAD
F-1800.0005BA F-1800.0025BA
DL-α-Methylleucine
DL-alpha-Methylleucine

Sequence: H-α-Me-DL-Leu-OH


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₇H₁₅NO₂
MW: 145.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038403
CAS Number: 144-24-1
EINECS: 205-621-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR