α-Methyl-DL-histidine dihydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-α-Me-DL-His-OH·2 HCl

4004262.0250 4004262.1000
F-1795.0250BAEA 309.81 CAD
F-1795.0250BA F-1795.1000BA
α-Methyl-DL-histidine dihydrochloride
alpha-Methyl-DL-histidine dihydrochloride

Sequence: H-α-Me-DL-His-OH · 2 HCl


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


αMeHis, racemate.

Formula: C₇H₁₃Cl₂N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00055953
CAS Number: 32381-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR