(R)-2-(Methylamino)-3-phenylpropanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: N-Methyl-D-phenylalanine, N-Me-D-Phe-OH

4009534.0001 4009534.0005
F-1785.0001BAEA 413.58 CAD
F-1785.0001BA F-1785.0005BA
(R)-2-(Methylamino)-3-phenylpropanoic acid
(R)-2-(Methylamino)-3-phenylpropanoic acid

Sequence: N-Me-D-Phe-OH


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
Short N-methyl-phenylalanine-rich peptides were shown to function as versatile blood-brain barrier shuttles.

Formula: C₁₀H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00069560
CAS Number: 56564-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR