2-(Methylamino)isobutyric acid hydrate
Supplier: BACHEM AMERICAS INC
Synonyms:
N,2-Dimethylalanine
4003409.0001
4003409.0005
F-1765.0001BAEA
206.51
CAD
F-1765.0001BA
F-1765.0005BA
2-(Methylamino)isobutyric acid hydrate
2-(Methylamino)isobutyric acid hydrate
Sequence: N-Me-Aib-OH
Synonym(s): N-Me-α-Me-Ala-OH
A non-metabolizable substrate for the amino acid transport system A. Used as amine component for studying difficult couplings.
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004191 CAS Number: 2566-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...