D(-)-Lysine monohydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: (R)-(-)-Lysine monohydrochloride, H-D-Lys-OH.HCl, D-Lysine hydrochloride, (R)-(-)-2,6-Diaminocaproic acid monohydrochloride, (R)-(-)-2,6-Diaminohexanoic acid monohydrochloride

4017512.0005 4017512.0025 4017512.0100
F-1730.0005BAEA 227.28 CAD
F-1730.0005BA F-1730.0025BA F-1730.0100BA
D(-)-Lysine monohydrochloride
D(-)-Lysine monohydrochloride

Sequence: H-D-Lys-OH · HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


[923-27-3] (net)

Formula: C₆H₁₅ClN₂O₂
MW: 182.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012920
CAS Number: 7274-88-6
EINECS: 230-691-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR