D-(-)-Leucine

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Leu-OH, (R)-(-)-Leucine

4007218.0005 4007218.0100
F-1700.0005BAEA 175.66 CAD
F-1700.0005BA F-1700.0100BA
D-(-)-Leucine
D(-)-Leucine

Sequence: H-D-Leu-OH


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₆H₁₃NO₂
Storage Temperature: Freezer
MDL Number: MFCD00063088
CAS Number: 328-38-1
EINECS: 206-327-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR