DL-Isoserine

Supplier: BACHEM AMERICAS INC

4006366.0001 4006366.0005 4006366.0025
F-1695.0001BAEA 185.46 CAD
F-1695.0001BA F-1695.0005BA F-1695.0025BA
DL-Isoserine
DL-Isoserine

Sequence: H-DL-Isoser-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.


Synonym(s): H-DL-β-Amino-α-hydroxypropionic acid

Formula: C₃H₇NO₃
Storage Temperature: Freezer
MDL Number: MFCD00008138
CAS Number: 565-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR