D-(-)-Isoleucine
Supplier: BACHEM AMERICAS INC
Synonyms:
(2R,3R)-(-)-Isoleucine, H-D-Ile-OH
4006515.0001
4006515.0005
F-1685.0001BAEA
627.31
CAD
F-1685.0001BA
F-1685.0005BA
D-(-)-Isoleucine
D(-)-Isoleucine
Sequence: H-D-Ile-OH
Formula:
H₃CCH₂CH(CH₃)CH(NH₂)COOH Storage Temperature: Ambient |
MDL Number:
MFCD00064221 CAS Number: 319-78-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...