L-Isoglutamine

Supplier: BACHEM AMERICAS INC

4001132.0001 4001132.0005
F-1680.0001BAEA 258.14 CAD
F-1680.0001BA F-1680.0005BA
L-Isoglutamine
L-Isoglutamine

Sequence: H-Glu-NH₂


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Synonym(s): H-Isogln-OH#L-Isoglutamine

Formula: C₅H₁₀N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00135661
CAS Number: 636-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR