4-Iodo-L-phenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: H-Phe(4-I)-OH

4001611.0001 4001611.0005
F-1665.0001BAEA 180.91 CAD
F-1665.0001BA F-1665.0005BA
4-Iodo-L-phenylalanine
4-Iodo-L-phenylalanine

Sequence: H-p-Iodo-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.


Educt for synthesizing Ar-modified phenylalanines by Suzuki-Miyaura coupling. p-Iodo-L-phenylalanine has been incorporated site-specifically into proteins for structure determination.

Formula: C₉H₁₀INO₂
Storage Temperature: Ambient
MDL Number: MFCD00002602
CAS Number: 24250-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR