(2S,4R)-Methyl-4-hydroxypyrrolidine-2-carboxylate hydrochloride

Supplier: BACHEM AMERICAS INC

4002341.0005 4002341.0025
F-1660.0005BAEA 180.91 CAD
F-1660.0005BA F-1660.0025BA
(2S,4R)-Methyl-4-hydroxypyrrolidine-2-carboxylate hydrochloride
(2S,4R)-Methyl-4-hydroxypyrrolidine-2-carboxylate hydrochloride

Sequence: H-Hyp-OMe · HCl


Hyp denotes L-trans-hydroxyproline.

Formula: C₆H₁₁NO₃·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00080855
CAS Number: 40216-83-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR