trans-4-Hydroxy-L(-)-proline

Supplier: BACHEM AMERICAS INC

4002357.0025 4002357.0100 4002357.0250
F-1655.0025BAEA 180.91 CAD
F-1655.0025BA F-1655.0100BA F-1655.0250BA
trans-4-Hydroxy-L(-)-proline
trans-4-Hydroxy-L(-)-proline

Sequence: H-Hyp-OH


Hyp denotes L-trans-hydroxyproline.


Synonym(s): (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid

Formula: C₅H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00064320
CAS Number: 51-35-4
EINECS: 200-091-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR