2,6-Diamino-5-hydroxyhexanoic acid hydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
H-DL-δ-Hydroxy-DL-Lys-OH·HCl
4002102.0001
4002102.0005
F-1650.0001BAEA
180.91
CAD
F-1650.0001BA
F-1650.0005BA
2,6-Diamino-5-hydroxyhexanoic acid hydrochloride
2,6-Diamino-5-hydroxyhexanoic acid hydrochloride
Sequence: H-DL-δ-Hydroxy-DL-Lys-OH · HCl
For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Synonym(s): H-DL-Hyl-OH · HCl
Formula:
C₆H₁₅ClN₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00012878 CAS Number: 13204-98-3 EINECS: 236-168-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...