(R)-(-)-2-Hydroxyisovaleric acid

Supplier: BACHEM AMERICAS INC

Synonyms: D-α-Hydroxyisovaleric acid, 2-Hydroxyisovaleric-acid, (R)-2-Hydroxy-3-methylbutanoic acid, D-Valic acid, (2R)-2-Hydroxy-3-methylbutanoic acid

4001785.0001 4001785.0005
F-1645.0001BAEA 264.44 CAD
F-1645.0001BA F-1645.0005BA
(R)-(-)-2-Hydroxyisovaleric acid
(R)-(-)-2-Hydroxyisovaleric acid

Sequence: D-α-Hydroxyisovaleric acid


Useful chiral synthons, building blocks for depsipeptides.


Synonym(s): D-Valic acid

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00066442
CAS Number: 17407-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR