(S)-(+)-2-Hydroxyisovaleric acid
Supplier: BACHEM AMERICAS INC
4001783.0005
4001783.0025
F-1640.0005BAEA
258.09
CAD
F-1640.0005BA
F-1640.0025BA
(S)-(+)-2-Hydroxyisovaleric acid
(S)-(+)-2-Hydroxyisovaleric acid
Sequence: L-α-Hydroxyisovaleric acid
Useful chiral synthons, building blocks for depsipeptides.
Synonym(s): L-Valic acid
Formula:
C₅H₁₀O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00066443 CAS Number: 17407-55-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...