L-Pipecolic acid

Supplier: BACHEM AMERICAS INC

4006540.0001 4006540.0005 4006540.0025
F-1625.0001BAEA 185.46 CAD
F-1625.0001BA F-1625.0005BA F-1625.0025BA
L-Pipecolic acid
L-Pipecolic acid

Sequence: H-Homopro-OH


Pip (or Hpr) derivatives.


Synonym(s): L-Pipecolic acid#H-Pip-OH

Formula: C₆H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00005981
CAS Number: 3105-95-1
EINECS: 221-462-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR