D-Homophenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: (R)-2-Amino-4-phenylbutyric Acid, H-D-HomoPhe-OH, H-D-HoPhe-OH

4005380.0001 4005380.0005 4005380.0025
F-1615.0001BAEA 180.91 CAD
F-1615.0001BA F-1615.0005BA F-1615.0025BA
D-Homophenylalanine
D-Homophenylalanine

Sequence: H-D-Homophe-OH


Hph derivatives. Bachem also offers Fmoc-Hph-Wang resin, D-2005.


Synonym(s): (R)-2-Amino-4-phenyl-butyric acid

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063091
CAS Number: 82795-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR