L-Histidinol dihydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: L-Histidinol·2 HCl, (S)-2-Amino-3-(1H-imidazol-4-yl)propan-1-ol dihydrochloride

4013484.0005 4013484.0025
F-1605.0005BAEA 635.29 CAD
F-1605.0005BA F-1605.0025BA
L-Histidinol dihydrochloride
L-Histidinol dihydrochloride

Sequence: L-Histidinol · 2 HCl


Reversible inhibitor of protein synthesis in cultured human cells. L-Histidinol was evaluated in cancer chemotherapy as an adjunctive enhancing the efficacy of anticancer drugs.

Formula: C₆H₁₃Cl₂N₃O
MW: 214.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00078058
CAS Number: 1596-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR