D-(-)-Glutamine

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Gln-OH, (R)-2,5-Diamino-5-oxopentanoic acid, (R)-(-)-Glutamine

4007503.0001 4007503.0005 4007503.0025
F-1580.0001BAEA 175.66 CAD
F-1580.0001BA F-1580.0005BA F-1580.0025BA
D-(-)-Glutamine
D(-)-Glutamine

Sequence: H-D-Gln-OH


For isoglutamine derivatives please see the family "Glutamic Acid".

Formula: C₅H₁₀N₂O₃
MW: 146.15
Storage Temperature: Refrigerator
MDL Number: MFCD00065607
CAS Number: 5959-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR