1-Benzyl-D-glutamate

Supplier: BACHEM AMERICAS INC

4004418.0001 4004418.0005
F-1555.0001BAEA 232.35 CAD
F-1555.0001BA F-1555.0005BA
1-Benzyl-D-glutamate
1-Benzyl-D-glutamate

Sequence: H-D-Glu-OBzl


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₂H₁₅NO₄
Storage Temperature: Freezer
MDL Number: MFCD00237065
CAS Number: 79338-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR