H-DL-β-(3,4-Dihydroxyphenyl)-DL-Ser-OH

Supplier: BACHEM AMERICAS INC

4006570.0050 4006570.0250 4006570.1000
F-1525.0050BAEA 206.51 CAD
F-1525.0050BA F-1525.0250BA F-1525.1000BA
H-DL-β-(3,4-Dihydroxyphenyl)-DL-Ser-OH
H-DL-β-(3,4-Dihydroxyphenyl)-DL-Ser-OH

Sequence: H-DL-β-(3,4-Dihydroxyphenyl)-DL-Ser-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.


Freeman et al. could show that DL-DOPS improves features of neurogenic orthostatic hypotension in patients with central and peripheral autonomic nervous system disease.

Formula: C₉H₁₁NO₅
Storage Temperature: Freezer
MDL Number: MFCD00004266
CAS Number: 3916-18-5
EINECS: 223-480-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR