2,6-Diaminopimelic acid (±) + meso

Supplier: BACHEM AMERICAS INC

4004244.0005 4004244.0001
F-1505.0005BAEA 413.58 CAD
F-1505.0005BA F-1505.0001BA
2,6-Diaminopimelic acid (±) + meso
2,6-Diaminopimelic acid

Mixture of LL-, DD- and meso-isomers

Formula: C₇H₁₄N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00002637
CAS Number: 583-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR