H-3,4-DehydroPro-OMe hydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
H-3,4-Dehydro-Pro-OMe·HCl
4004048.0250
4004048.1000
F-1500.0250BAEA
335.58
CAD
F-1500.0250BA
F-1500.1000BA
H-3,4-DehydroPro-OMe hydrochloride
H-3,4-DehydroPro-OMe hydrochloride
Sequence: H-3,4-Dehydro-Pro-OMe · HCl
Dhp derivatives.
Formula:
C₆H₁₀ClNO₂ MW: 163.601 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00038999 CAS Number: 186145-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...