3,4-Dehydro-L-proline

Supplier: BACHEM AMERICAS INC

4003545.0250 4003545.1000
F-1490.0250BAEA 593.81 CAD
F-1490.0250BA F-1490.1000BA
3,4-Dehydro-L-proline
3,4-Dehydro-L-proline

Sequence: H-3,4-Dehydro-Pro-OH


Dhp derivatives.

Formula: C₅H₇NO₂
Storage Temperature: Freezer
MDL Number: MFCD00065964
CAS Number: 4043-88-3
EINECS: 223-738-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR