4-Chloro-DL-phenylalanine

Supplier: BACHEM AMERICAS INC

4002988.0025 4002988.0100
F-1450.0025BAEA 304.06 CAD
F-1450.0025BA F-1450.0100BA
4-Chloro-DL-phenylalanine
4-Chloro-DL-phenylalanine

Sequence: H-p-Chloro-DL-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.


Synonym(s): (RS)-2-Amino-3-(4-chloro-phenyl)-propionic acid#H-4-Chloro-DL-Phe-OH#Fenclonine#PCPA

Formula: C₉H₁₀ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00002601
CAS Number: 7424-00-2
EINECS: 231-051-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR