4-Chloro-L-phenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: L-4-Chlorophenylalanine, H-Phe(4-Cl)-OH

4002990.0001 4002990.0005
F-1445.0001BAEA 206.51 CAD
F-1445.0001BA F-1445.0005BA
4-Chloro-L-phenylalanine
4-Chloro-L-phenylalanine

Sequence: H-4-Chloro-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.


Synonym(s): (S)-2-Amino-3-(4-chloro-phenyl)-propionic acid#H-4Cpa-OH#L-Fenclonine#L-PCPA#H-p-Chloro-Phe-OH

Formula: C₉H₁₀ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00063065
CAS Number: 14173-39-8
EINECS: 238-023-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR