3-Chloro-L-alanine hydrochloride

Supplier: BACHEM AMERICAS INC

4000633.0001 4000633.0005
F-1430.0001BAEA 180.91 CAD
F-1430.0001BA F-1430.0005BA
3-Chloro-L-alanine hydrochloride
3-Chloro-L-alanine hydrochloride

Sequence: H-β-Chloro-Ala-OH · HCl


Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines


Non-competitive inhibitor of alanine racemase, Ki 1.71 mM.

Formula: C₃H₆ClNO₂
Storage Temperature: Freezer
MDL Number: MFCD00070398
CAS Number: 51887-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR