(R)-2-Amino-5-(((benzyloxy)carbonyl)amino)pentanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Orn(Z)-OH

4010077.0005 4010077.0025
F-1405.0005BAEA 619.6 CAD
F-1405.0005BA F-1405.0025BA
(R)-2-Amino-5-(((benzyloxy)carbonyl)amino)pentanoic acid
(R)-2-Amino-5-(((benzyloxy)carbonyl)amino)pentanoic acid

Sequence: H-D-Orn(Z)-OH

Formula: C₁₃H₁₈N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00063115
CAS Number: 16937-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR