H-D-Lys(Z)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Lys(Cbz)-OH, Nepsilon-Benzyloxycarbonyl-D-lysine, Nepsilon-Cbz-D-lysine, H-D-Lys(Z)-OH, Nepsilon-Carbobenzoxy-D-lysine

4003804.0005 4003804.0025
F-1400.0005BAEA 258.14 CAD
F-1400.0005BA F-1400.0025BA
H-D-Lys(Z)-OH
H-D-Lys(Z)-OH

Sequence: H-D-Lys(Z)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₄H₂₀N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00063120
CAS Number: 34404-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR