cis-4-Hydroxy-D(+)-proline

Supplier: BACHEM AMERICAS INC

Synonyms: (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline

4010450.0001 4010450.0005 4010450.0025
F-1395.0001BAEA 180.91 CAD
F-1395.0001BA F-1395.0005BA F-1395.0025BA
cis-4-Hydroxy-D(+)-proline
cis-4-Hydroxy-D(+)-proline

Sequence: H-D-cis-Hyp-OH


Hyp denotes L-trans-hydroxyproline.


Synonym(s): (2R,4R)-4-Hydroxypyrrolidine-2-carboxylic acid

Formula: C₅H₉NO₃
MW: 131.13
Storage Temperature: Ambient
MDL Number: MFCD00005252
CAS Number: 2584-71-6
EINECS: 219-963-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR