γ-Carboxyglutamic acid

Supplier: BACHEM AMERICAS INC

Synonyms: H-γ-Carboxy-Glu-OH

4001771.0010 4001771.0050 4001771.0250
F-1380.0010BAEA 284.04 CAD
F-1380.0010BA F-1380.0050BA F-1380.0250BA
γ-Carboxyglutamic acid
gamma-Carboxyglutamic acid

Sequence: H-γ-Carboxy-Glu-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Synonym(s): H-Gla-OH

Formula: C₆H₉NO₆
Storage Temperature: Freezer
MDL Number: MFCD00011673
CAS Number: 53861-57-7
EINECS: 258-825-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR