3-Ureidopropionic acid

Supplier: BACHEM AMERICAS INC

4017082.0005 4017082.0001
F-1375.0005BAEA 722.85 CAD
F-1375.0005BA F-1375.0001BA
3-Ureidopropionic acid
3-Ureidopropionic acid

Carbamoyl-b-Ala-OH
CAS: 462-88-4
Formula: C4H8N2O3
M.W: 132.12
Storage temp.: RT

Formula: C₄H₈N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00043023
CAS Number: 462-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR