(R)-2-Amino-4,4-dimethylpentanoic acid

Supplier: BACHEM AMERICAS INC

4019075.0001 4019075.0005
F-1320.0001BAEA 672.04 CAD
F-1320.0001BA F-1320.0005BA
(R)-2-Amino-4,4-dimethylpentanoic acid
(R)-2-Amino-4,4-dimethylpentanoic acid

Sequence: H-β-tBu-D-Ala-OH


Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
γ-Methylleucine, also called neopentylglycine.


Synonym(s): H-D-Neopentylgly-OH#H-γ-Me-D-Leu-OH#H-D-Tba-OH

Formula: C₇H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00038404
CAS Number: 88319-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR