4-Bromo-L-phenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: H-Phe(4-Br)-OH

4007011.0001 4007011.0005
F-1305.0001BAEA 284.04 CAD
F-1305.0001BA F-1305.0005BA
4-Bromo-L-phenylalanine
4-Bromo-L-phenylalanine

Sequence: H-p-Bromo-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.

Formula: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD00063062
CAS Number: 24250-84-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR