DL-2-Aminopimelic acid

Supplier: BACHEM AMERICAS INC

Synonyms: DL-2-Aminoheptanedioic acid, 2-Aminoheptanedioic acid

4014992.0001 4014992.0005 4014992.0025
F-1235.0001BAEA 185.3 CAD
F-1235.0001BA F-1235.0005BA F-1235.0025BA
DL-2-Aminopimelic acid
DL-2-Aminopimelic acid

Sequence: DL-2-Aminoheptanedioic acid
Synonym(s): DL-α-Aminopimelic acid#H-DL-Apm-OH


Peptides containing 2-aminopimelic acid (Apm) inhibit diaminopimelic acid biosynthesis in bacteria.

Formula: C₇H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00053314
CAS Number: 627-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR